ST-200-MP Stuart Three Position Magnetic Stirrer
ST-200-MP Stuart Three Position Magnetic Stirrer
Actual product may vary
Fisherbrand
ST-200-MP Stuart Three Position Magnetic Stirrer
ST-200-MP Stuart Three Position Magnetic Stirrer has three independent stirring positions yet only measures 23-5/8 in. L x 10-5/8 in. W (60 x 27 cm).
Have Questions?
Catalog NumberModel
FKS11426538SB161-3
Catalog number FKS11426538
Price (SGD)
4,474.00
EA
Add to cart
Model:
SB161-3
Price (SGD)
4,474.00
EA
Add to cart
  • Powerful magnets for strong magnetic coupling and stirring capacity
  • Easily accommodate a 2-L beaker in each position
  • Stainless steel top plate does not produce eddy current to ensure powerful stirring action
Three independently controlled position units only require one power source. Run three separate tests at the same time or run multiple samples under the same conditions.
Specifications
Speed100 to 1500 rpm
Stirring Positions3
Voltage230 V
Wattage150 W
Dimensions (L x W)550 x 210 mm
Dimensions (L x W x H)60 x 27 x 11 cm
Height (English)4.3 in.
Height (Metric)11 cm
Length (English)23.6 in.
Length (Metric)60 cm
Width (English)10.6 in.
Width (Metric)27 cm
Electrical Requirements230 V, 50 Hz, 150 W
Frequency50 Hz
Certifications/ComplianceEN 61326-1:2006, EN 61010-1:2001, EN 61010-2-051:2003
Includes3 x 25 mm PTFE Coated Stir Bars
ModelSB161-3
TypeMagnetic Stirrer
Weight (English)15.4 lb.
Weight (Metric)7 kg
Unit SizeEA