Stuart™ Analogue Tube Roller
Stuart™ Analogue Tube Roller
Actual product may vary
Fisherbrand
Stuart™ Analogue Tube Roller
Provides a gentle, but highly efficient, rocking and rolling action. Stuart™ Analogue Tube Roller Units can be used in incubators up to 60°C and humidity up to 80%, or in cold rooms down to 4°C.
Have Questions?
Catalog NumberIncludesDepth (Metric)
FKS114164989 Rollers375 mm
Catalog number FKS11416498
Price (SGD)
4,948.00
EA
Add to cart
Includes:
9 Rollers
Depth (Metric):
375 mm
Price (SGD)
4,948.00
EA
Add to cart
Provides a gentle, but highly efficient, rocking and rolling action. The six rollers design has a small space-saving footprint, while the nine roller unit has a higher capacity.
  • Control is through a simple on/off switch and the unit will run continuously at the fixed speed of 33rpm
  • Recommended for mixing blood samples, viscous substances and liquid-solid suspensions where minimum aeration is required.
  • Units can be used in incubators up to 60°C and humidity up to 80%, or in cold rooms down to 4°C. Both roller mixers are robustly constructed and designed for easy cleaning, having plastic rollers and a drip tray to collect accidental spillages.
  • Most sizes of tubes, Bijoux, Universals and bottles can be accommodated, up to 100mm diameter.
Specifications
Speed Range33 rpm
Electrical Requirements230 V, 50 Hz, 50 W
Voltage230 V
Wattage50 W
Frequency50 Hz
Depth (Metric)375 mm
Height (Metric)110 mm
Length (Metric)340 mm
Width (Metric)595 mm
Controller TypeAnalogue
ModelSRT9
For Use With (Application)Mixing blood samples, viscous substances and liquid-solid suspensions where minimum aeration is required
Humidity RangeUp to 80%
Includes9 Rollers
Packaging Quantity1 Ea.
Unit SizeEA