Check Order Status
Stainless Steel Gabled Lid
Actual product may vary
Fisherbrand
Stainless Steel Gabled Lid
For use with OLS Aqua Pro Orbital and Linear Shaking bath
Have Questions?
Catalog number
FKS11400948
Price (SGD)
1,272.00
EA
Add to cart
Price (SGD)
1,272.00
EA
Add to cart
For use with OLS Aqua Pro Orbital and Linear Shaking bath
Gabled
Stainless steel construction
Includes access hole for cooling accessories
For use with OLS Aqua Pro
Specifications
For Use With (Equipment)
OLS 200 Shaking Water Bath
Shaking Motion
Linear