Polypropylene Pipette Racks
Polypropylene Pipette Racks
Actual product may vary
Polypropylene Pipette Racks
Rotating base holds up to 94 pipettes. Kartell™ Polypropylene Pipette Racks safely and conveniently store pipettes of various sizes.
Have Questions?
Catalog NumberPositions
FKS1139837094
Catalog number FKS11398370
Price (SGD)
200.00
EA
Add to cart
Positions:
94
Price (SGD)
200.00
EA
Add to cart
  • Desktop holder keeps pipettes organized and within reach
  • Swivel two tiered stand holds pipettes with a diameter up to 14mm
  • Polypropylene construction for durability
Specifications
Product TypePipet Stand
HoldsPipettes (94)
MaterialPolypropylene
Positions94
For Use With (Equipment)Desktop Pipette Holder
No. per Pack1 Pack
Quantity1 Pack
Height (Metric)470 mm
Diameter (Metric) Outer230 mm
Unit SizeEA