Class A Reusable PFA Volumetric Flasks with Screw Cap
Class A Reusable PFA Volumetric Flasks with Screw Cap
Actual product may vary
Fisher Catalogue
Class A Reusable PFA Volumetric Flasks with Screw Cap
Autoclaved to 121°C
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
FKS11397881100 mL
FIS10-210-64410 mL
FKS1137788110 mL
FIS10-210-64525 mL
FKS1138788125 mL
FIS10-210-64650 mL
FKS1131789150 mL
FIS10-210-649500 mL
Catalog number FKS11397881
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity (Metric):
100 mL
Translucent flask is supplied with a perfluoroalkoxy-copolymer screw cap. Screw caps provide hermetic seals and help reduce the possibility of outside contamination. The product is manufactured to Class A tolerances as defined by ISO 1042 and flasks have individually calibrated graduation lines. They withstand high temperatures and are resistant to a variety of aqueous solutions and organic solvents. These flasks may be autoclaved to 121°C without affecting the graduation tolerance. Recommended routine cleaning up to 60°C will preserve graduations. Height dimension is flask only, without screw cap.
Specifications
Capacity (English)3.38 oz.
Closure MaterialPerfluoroalkoxy Copolymer
ColorTranslucent
Diameter (English) Outer2.4 in.
Capacity (Metric)100 mL
MaterialPFA
Height (English)6.4 in.
Height (Metric)16.5 cm
Temperature Range (English)140°F
Temperature Range (Metric)60°C
Tolerance0.1 mL
AutoclavableAutoclavable
DisposableReusable
Bottom ShapeFlat
Diameter (Metric) Outer6.09 cm
Closure SizeGL18
Unit SizePK