Whatman™ Polyethersulfone (PES) Membrane Filters
Whatman™ Polyethersulfone (PES) Membrane Filters
Actual product may vary
Fisherbrand
Whatman™ Polyethersulfone (PES) Membrane Filters
Whatman polyethersulfone (PES) membrane filters from Cytiva’s Life Sciences business are hydrophilic and stable in alkaline pH, making them suitable for biological samples and aqueous applications.
Have Questions?
Catalog NumberPore Size
FKS113889940.8 μm
Catalog number FKS11388994
Price (SGD)
-
Pore Size:
0.8 μm
  • Low protein binding minimizes nonspecific protein adsorption
  • Hydrophilic membrane suitable for aqueous applications, especially in alkaline pH
  • Smooth surface for easy enumeration of microorganisms
Specifications
Pore Size0.8 μm
Diameter (Metric)47 mm
Unit SizePK