1x1 image pixel for data collection
Whatman™ Regenerated Cellulose Membranes: Grade RC 58
Whatman™ Regenerated Cellulose Membranes: Grade RC 58
Actual product may vary
Fisherbrand
Whatman™ Regenerated Cellulose Membranes: Grade RC 58
Consists of pure cellulose without any wetting agents
Have Questions?
Catalog NumberDiameter (Metric)DimensionsLxW
FKS1138539447 mm
Catalog number FKS11385394
Price (SGD)
1,376.00
PK
Estimated availability date 01-Oct-2025
Add to cart
Diameter (Metric):
47 mm
Price (SGD)
1,376.00
PK
Add to cart
Consists of pure cellulose without any wetting agents
  • Spontaneously wetting, very good wet strength
  • Extremely chemically resistant; suitable for aqueous and organic media
  • Hydrophilic
  • Mechanically stable
  • Sterilizable by all methods
Specifications
Filter TypeMembrane Filter
MaterialRegenerated Cellulose
Pore Size0.2 μm
SterilitySterile
HydrophobicityHydrophilic
Membrane TypeRC58
Thickness (Metric)75 μm
TypeFiltration Membrane Disc
Diameter (Metric)47 mm
Unit SizePK
Share catalog number, name or link