Micro Haematocrit Capillaries, CE-IVD
Micro Haematocrit Capillaries, CE-IVD
Actual product may vary
Fisherbrand
Micro Haematocrit Capillaries, CE-IVD
Heparinised with sodium heparinate over internal surface. BRAND™ Micro-haematocrit Capillary Tubes coded red for capillary blood. Disposable without calibration mark.
Have Questions?
Catalog NumberAdditive
FKS11383994Sodium Heparin
Catalog number FKS11383994
Price (SGD)
143.00
PK
Add to cart
Additive:
Sodium Heparin
Price (SGD)
143.00
PK
Add to cart
  • DIN ISO 12772 and BS 4316-68
  • With color code
  • For use in haematocrit centrifuges. 
  • Heparinised: Heparinised with sodium heparinate over internal surface, red color code, for capillary blood.
  • Non-heparinised: Blue color code, for use with heparinised venous blood.
  • CE marked according to guideline EU IVDR 2017/746
Specifications
AdditiveSodium Heparin
Diameter (Metric) Inner1.15 ± 0.05 mm
For Use With (Equipment)Hematocrit Centrifuges
Closure ColorRed
For Use With (Application)For use with heparinized capillary blood
Length (Metric)75 ± 0.5 mm
Unit SizePK