Azlon™ Polypropylene Slide Staining Jar
Azlon™ Polypropylene Slide Staining Jar
Actual product may vary
Fisherbrand
Azlon™ Polypropylene Slide Staining Jar
Useful for staining, biochemical tests, small batches or for mailing of slides
Have Questions?
Catalog NumberDiameter (Metric)
FKS1138369433 mm
Catalog number FKS11383694
Price (SGD)
-
Diameter (Metric):
33 mm
  • Robust polypropylene jar
  • Can hold 10 slides back to back
  • Design conserves space and staining solution
  • Useful for staining, biochemical tests, small batches or for mailing of slides
Specifications
TypeSlide Staining Jar
AutoclavableAutoclavable
Capacity (Metric)10
ColorTranslucent
Diameter (Metric)33 mm
For Use With (Application)For staining, biochemical tests, small batches or for mailing of slides
Height (English)3.26 in.
Height (Metric)83 mm