Whatman™ Grade 1 Chr Cellulose Chromatography Paper
Whatman™ Grade 1 Chr Cellulose Chromatography Paper
Actual product may vary
Fisherbrand
Whatman™ Grade 1 Chr Cellulose Chromatography Paper
Ensures the wicking capability and uniformity of capillary action that are important in chemical separations. Whatman™ Grade 1 Chr Cellulose Chromatography paper is a smooth surface, 0.18 mm thick paper with a linear flow rate (water) of 130 mm/30 min.
Have Questions?
Catalog NumberLength (Metric)Width (Metric)Format
FKS1138072430 cm10 cmSheet
Catalog number FKS11380724
Price (SGD)
283.00
PK
Add to cart
Length (Metric):
30 cm
Width (Metric):
10 cm
Format:
Sheet
Price (SGD)
283.00
PK
Add to cart
Ensures the wicking capability and uniformity of capillary action that are important in chemical separations. Whatman™ Grade 1 Chr Cellulose Chromatography paper is a smooth surface, 0.18 mm thick paper with a linear flow rate (water) of 130 mm/30 min.
The world standard chromatography paper: 18mm paper for general analytical separations, available in rolls, sheets or strips.
  • Pure cellulose produced entirely from the highest quality cotton linters with no additives of any kind.
  • Manufactured and tested specifically for chromatographic techniques.
  • Widely used in protein and nucleic acid blotting.
  • Good resolution for general analytical separations.
Specifications
Description1 Chr cellulose chromatography roll is a 0.18 mm paper for general analytical separations.
Dimensions (L x W)10 x 30 cm (3.93 x 11.8 in.)
Film Thickness0.18 mm
FinishSmooth
For Use With (Application)General Analytical Separations, Protein and Nucleic Acid Blotting
FormatSheet
GradeGrade 1 Chr
Length (English)11.8 in.
MaterialHigh-quality Cotton Linters
Width (English)3.93 in.
Width (Metric)10 cm
Flow Rate130 mm/30 min.
Length (Metric)30 cm
Unit SizePK