XK Chromatography Columns (Empty)
XK Chromatography Columns (Empty)
Actual product may vary
Fisher Catalogue
XK Chromatography Columns (Empty)
Ensure trouble-free operation for low to medium pressure. Cytiva XK Chromatography Columns (Empty) connect directly to ÄKTA design instruments or other liquid chromatography systems. The user-friendly ensures reproducible, accurate packing.
Have Questions?
Catalog NumberHeight (Metric)Diameter (Metric) Inner
FKS11380332400 mm26 mm
Catalog number FKS11380332
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Height (Metric):
400 mm
Diameter (Metric) Inner:
26 mm
XK columns are constructed of robust borosilicate glass, which exhibits excellent chemical resistance and is compatible with aqueous solutions and most organic solvents used in liquid chromatography of macromolecules. The clear tubes allow for visual inspection of the media bed and are available in a broad range of column dimensions for use with many different purification needs.
Column packing can be performed using either a packing reservoir or an extra column tube attached with a packing connector. The QuickLock mechanism of the adapter shaft facilitates rapid and easy movement of the adapter, simplifying adjustments of the bed height, disassembly, and cleaning. The design of the adapter plunger gives a uniform flow, which maintains the integrity of the packed bed during operations.
  • Plunger design to ensure a homogeneous buffer distribution and reproducible packing
  • Adapter QuickLock mechanism that facilitates column handling and cleaning
  • Fittings for direct connection to chromatographiy systems
  • Cooling jacket for sensitive purifications
  • Extensive range of spare parts available, facilitating maintenance
Not resistant to Acetone, Ketones, Chlorinated Hydrocarbons, Aliphatic Esters and Phenol.
Should be avoided: >10% NaOH, >10% HCl, >5% acetic acid, and strong mineral acids.
Specifications
Product TypeXK Chromatography Columns (Empty)
Bed VolumeBed height (one adapter): 230 to 350mm
Bed height (two adapters): 85 to 350mm
Diameter (Metric) Inner26 mm
Height (Metric)400 mm
Flow RateInlet/Outlet (i.d.): 0.75mm
MaterialBorosilicate glass
Max. Pressure5 bar
Particle Size10μm pore size
Support TypePolypropylene (reinforced glass fiber)/Polyamide Net
Column TypeEmpty Chromatography Column
For Use With (Application)Liquid chromatography at low to medium pressure
Unit SizePK