Sterilin™ 55mm Contact Plate
Sterilin™ 55mm Contact Plate
Actual product may vary
Sterilin™ 55mm Contact Plate
Sterilin™ 55mm Contact Plate
Sterilin™ 55mm Contact Plate
Fisherbrand
Sterilin™ 55mm Contact Plate
Perform routine hygiene monitoring of surfaces using Thermo Scientific™ Sterilin™ 55mm Contact Plate Petri Dishes.
Have Questions?
Catalog NumberDiameter (Metric)
FKS1137927367 mm
Catalog number FKS11379273
Price (SGD)
1,463.00
PK
Add to cart
Diameter (Metric):
67 mm
Price (SGD)
1,463.00
PK
Add to cart
Perform routine hygiene monitoring of surfaces using Thermo Scientific™ Sterilin™ 55mm Contact Plate Petri Dishes.
NOT AVAILABLE IN AMERICA
Features:
  • Domed base aids media adhesion
  • Triple vented aids gaseous exchange
  • Numbered grid on the base facilitates colony counting
  • Deep skirted base aids stability when stacked
  • Aseptically manufactured
Ordering Information:
Not available in all geographic regions
Specifications
Certifications/Compliance25cm² surface area-conforms to both the Institute of Environmental Sciences (IES, 1993) and the International Pharmaceutical Federation (I.P.F., 1990) Standards
DescriptionContact Plate, 55mm
Culture Area25 sq. cm
ShapeRound
SterilityAseptically Manufactured
TypeContact Plate
No. per Case300
No. per Pack10
Vent3
Height (Metric)10.4 mm
Diameter (Metric)67 mm
Unit SizePK