Sterilin Autoclave Bags are strong disposable bags with blue biohazard printing specifically designed for contaminated waste disposal in autoclaves or incinerators.
Have Questions?
Catalog Number
Dimensions (L x W)
Material
No. per Case
FKS11379103
660 x 305mm
Polypropylene
200
Catalog number FKS11379103
Price (SGD)
628.00
PK
Add to cart
Dimensions (L x W):
660 x 305mm
Material:
Polypropylene
No. per Case:
200
Price (SGD)
628.00
PK
Add to cart
Sterilin Autoclave Bags are strong disposable bags with blue biohazard printing specifically designed for contaminated waste disposal in autoclaves or incinerators.
HDPE bags are suitable for sterilization at 121°C and polypropylene bags are suitable for the inactivation of particularly resistant biological waste at 135°C