Check Order Status
Moisture Test Paper
Actual product may vary
Fisherbrand
Moisture Test Paper
Moisture test paper for use when drying samples during moisture assessment
Have Questions?
Catalog Number
Diameter (Metric)
FKS11375124
90 mm
Catalog number
FKS11375124
Price (SGD)
300.00
PK
Add to cart
Diameter (Metric):
90 mm
Price (SGD)
300.00
PK
Add to cart
Used when drying samples during moisture assessement
Maximum recommended temperature - 550°C
Specifications
Material
Borosilicate Glass
For Use With (Application)
For use when drying samples during moisture assessment
Grade
Moisture Test Paper
Diameter (Metric)
90 mm