Disposable Polystyrene Cell Counter Cup
Disposable Polystyrene Cell Counter Cup
Actual product may vary
Fisher Catalogue
Disposable Polystyrene Cell Counter Cup
Manufactured with clear polystyrene for optimum visibility. Kartell™ Coulter™ Cell Counter Cups are used for specimen analysis with cell counters.
Have Questions?
Catalog NumberCapacity (Metric)
FKS1137380425 mL
Catalog number FKS11373804
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity (Metric):
25 mL
  • Disposable
  • Includes polyethylene caps
Specifications
Quantity200 Pk.
Capacity (Metric)25 mL
ColorClear
Product TypeCup
MaterialPolystyrene
DisposableSingle-use
For Use With (Application)For Analysis
For Use With (Equipment)Fr Toa™ and Royco-Hycel™ cell counters
Height (Metric)56 mm
Unit SizePK