Whatman™ Puradisc 25mm PTFE Syringe Filters
Whatman™ Puradisc 25mm PTFE Syringe Filters
Actual product may vary
Fisher Catalogue
Whatman™ Puradisc 25mm PTFE Syringe Filters
Contains hydrophobic membrane that is chemically stable and inert. Whatman™ Puradisc 25mm PTFE Syringe Filters exhibit low protein binding, and its main application is filtration of non-aqueous samples.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberPore SizeQuantity
FKS113733150.45 μm1000/Pk.
FIS05-713-3920.45 μm1000/Pk.
FKS113346840.1 μm50/Pk.
Catalog number FKS11373315
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Pore Size:
0.45 μm
Quantity:
1000/Pk.
Contains hydrophobic membrane that is chemically stable and inert. Whatman™ Puradisc 25mm PTFE Syringe Filters exhibit low protein binding, and its main application is filtration of non-aqueous samples.
  • For filtering solvents, chemicals and nonaqueous solutions
  • Typical applications include HPLC, TLC, and GC sample clarification; HPLC solvent filtration
  • Made of hydrophobic polytetrafluoroethylene (PTFE)
  • Nonsterile, autoclavable
Specifications
MaterialPTFE
Material (Membrane)PTFE
Pore Size0.45 μm
SterilityNon-sterile
Filtration Area4.2 cm2
For Use With (Application)For HPLC, TLC, and GC sample clarification; HPLC solvent filtration
Housing MaterialPigment free polypropylene (PP)
Inlet StyleFemale Luer Lock
Outlet StyleMale Luer
Pressure Rating5.2 bar (75 psi)
Quantity1000/Pk.
TypeSyringe Filter
Volume (Metric)Up to 100 mL
Dimensions (L x W x H)22.9 x 28.4 mm
Diameter (Metric)25 mm
Unit SizePK