Whatman™ Qualitative Filter Paper: Grade 113 Circles
Whatman™ Qualitative Filter Paper: Grade 113 Circles
Actual product may vary
Fisherbrand
Whatman™ Qualitative Filter Paper: Grade 113 Circles
Strengthened with chemically stable resin additive. Whatman™ Qualitative Filter Paper: Grade 113 Circles boast the fastest flow rate of the qualitative grades. Crepe surface promotes ultra high loading capacity.
Have Questions?
Catalog NumberDiameter (Metric)
FKS11372905125 mm
Catalog number FKS11372905
Price (SGD)
152.00
PK
Add to cart
Diameter (Metric):
125 mm
Price (SGD)
152.00
PK
Add to cart
Strengthened with chemically stable resin additive. Whatman™ Grade 113 Wet-Strengthened Qualitative Filter Paper Circles boast the fastest flow rate of the qualitative grades. Creped surface promotes ultra high loading capacity.
These extremely strong filter papers have a high wet strength due to the addition of a small quantity of chemically stable resin. Normal qualitative applications will not introduce any significant impurities into the filtrate.
  • Ultra high loading capacity with a particle retention well suited for use with coarse or gelatinous 
  • Fastest flow rate of the qualitative grades
  • Crepe surface
  • Thickest filter paper in the qualitative range
Note: Resins contain nitrogen so these grades should not be used in Kjeldahl estimations or similar operations.
Specifications
DescriptionGrade 113 Qualitative Filter Paper Wet-Strengthened, circle, 125 mm
FormatDisc
MaterialCellulose
Particle Retention30 μm
Filtration Speed1.3 s/100 mL/in2
For Use With (Application)Ultra high loading capacity with a particle retention well suited for use with coarse or gelatinous precipitates. Fastest flow rate of the qualitative grades. Creped surface. Thickest filter paper in the qualitative range.
GradeGrade 113
Quantity100 Pk.
Thickness (Metric)420 μm
TypeQualitative Filter Paper
Diameter (English)4.92 in.
Diameter (Metric)125 mm
Unit SizePK