Poxygrid™ 50 mL Centrifuge Tube Racks
Poxygrid™ 50 mL Centrifuge Tube Racks
Actual product may vary
Poxygrid™ 50 mL Centrifuge Tube Racks
Poxygrid™ 50 mL Centrifuge Tube Racks
Poxygrid™ 50 mL Centrifuge Tube Racks
Poxygrid™ 50 mL Centrifuge Tube Racks
Fisherbrand
Poxygrid™ 50 mL Centrifuge Tube Racks
Organizes tubes for storage and processing.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberPositionsDimensions (L x W x H)
FKS11372204814.9 x 8 x 9.2 cm
FISS967111614.9 x 15.6 x 9.2 cm
Catalog number FKS11372204
Price (SGD)
228.00
EA
Add to cart
Positions:
8
Dimensions (L x W x H):
14.9 x 8 x 9.2 cm
Price (SGD)
228.00
EA
Add to cart
  • Heat-cured epoxy coating is unaffected by organic solvents, salts, weak acids and weak alkalis
  • Steam autoclavable at 121 degree C (250 degree F)
  • Temperature range: -223° to 121°C (-369° to 250°F)
  • Supports 50 mL plastic centrifuge tubes in individual wells, each 30 mm square
Specifications
Product TypeCentrifuge Tube Rack
Array2 x 4
AutoclavableAutoclavable
ColorBlue
HoldsTubes (50 mL)
MaterialEpoxy-coated Steel
Positions8
ShapeRectangular
Shelves3 Tiers
StackableYes, When Empty
SterilityNon-sterile
Dimensions (L x W x H)14.9 x 8 x 9.2 cm
Length (Metric)14.9 cm
Length (English)5.87 in.
Width (Metric)8 cm
Width (English)3.18 in.
Height (Metric)9.2 cm
Height (English)3.62 in.
Unit SizeEA