Quickfit™ Borosilicate Glass Condenser Coil with Ground Cone and Socket
Quickfit™ Borosilicate Glass Condenser Coil with Ground Cone and Socket
Actual product may vary
Fisherbrand
Quickfit™ Borosilicate Glass Condenser Coil with Ground Cone and Socket
Supplied with detachable plastic screwthread connectors which allow 9mm flexible tubing to be fitted easily and safely. Quickfit™ Borosilicate Glass Condenser Coil with Ground Cone and Socket is a highly efficient condenser with a large cooling surface. Suitable for either reflux or distillation work.
Have Questions?
Catalog NumberStandard Taper Joint NoLength (Metric) Jacket
FKS1137185524/29465 mm (Overall) 320 mm (Effective)
Catalog number FKS11371855
Price (SGD)
649.00
EA
Add to cart
Standard Taper Joint No:
24/29
Length (Metric) Jacket:
465 mm (Overall) 320 mm (Effective)
Price (SGD)
649.00
EA
Add to cart
Supplied with detachable plastic screwthread connectors which allow 9mm flexible tubing to be fitted easily and safely. Quickfit™ Borosilicate Glass Condenser Coil with Ground Cone and Socket is a highly efficient condenser with a large cooling surface. Suitable for either reflux or distillation work.
  • Suitable for either reflux or distillation work
  • Highly efficient condensers with a large cooling surface
  • Supplied with detachable plastic screwthread connectors which allow 9mm flexible tubing to be fitted easily and safely
Specifications
For Use With (Application)Suitable for either reflux or distillation work
Length (Metric) Jacket465 mm (Overall) 320 mm (Effective)
Standard Taper Joint No24/29
MaterialBorosilicate Glass
TypeCondenser Coil
Unit SizeEA