PEEK Connector 1/16 in. Luer
PEEK Connector 1/16 in. Luer
Actual product may vary
Fisherbrand
PEEK Connector 1/16 in. Luer
Set of two (2) polyetheretherketone 1/16 in. Male/Luer Female connectors for connection of a syringe to the top of a chromatography column in the ÄKTA Chromatography System.
Have Questions?
Catalog NumberConnections
FKS113702321/16 in. Tapered Male to Luer Female
Catalog number FKS11370232
Price (SGD)
560.00
PK
Add to cart
Connections:
1/16 in. Tapered Male to Luer Female
Price (SGD)
560.00
PK
Add to cart
ÄKTA Laboratory-scale Systems are liquid chromatography systems used for protein purification at research laboratory scale.
  • Tapered syringe ends
  • Polyetheretherketone (PEEK) connstruction
  • Connections: 1/16 in. male–Luer female
Specifications
TypeConnector
Connections1/16 in. Tapered Male to Luer Female
QuantitySet of 2
MaterialPEEK
ColorBlack
For Use With (Equipment)ÄKTA Laboratory-Scale Systems, Syinge
Unit SizePK