Congo paper MN 260 HE pH Paper
Congo paper MN 260 HE pH Paper
Actual product may vary
Fisher Catalogue
Congo paper MN 260 HE pH Paper
Have Questions?
Catalog NumberColor Indicator
FKS11366514Red to Blue
Catalog number FKS11366514
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Color Indicator:
Red to Blue
Specifications
Color IndicatorRed to Blue
pH Range5 to 3
FormatStrip
For Use With (Application)Determination of lactic acid
Quantity200 Pack
TypepH Paper
Unit SizeEA