Whatman™ Vacu-Guard Filter
Whatman™ Vacu-Guard Filter
Actual product may vary
Whatman™ Vacu-Guard Filter
Whatman™ Vacu-Guard Filter
Fisherbrand
Whatman™ Vacu-Guard Filter
Helps to confine and isolate infectious materials in vacuum systems and protect the laboratory. Whatman™ Vacu-Guard Filter protects vacuum pump systems from solvent vapor or gaseous contaminants and aqueous aerosols.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDiameter (Metric) DiscFiltration Area
FKS1136463450 mm16 cm2
FIS09-744-7550 mm16 cm2
Catalog number FKS11364634
Price (SGD)
894.00
PK
Add to cart
Diameter (Metric) Disc:
50 mm
Filtration Area:
16 cm2
Price (SGD)
894.00
PK
Add to cart
Helps to confine and isolate infectious materials in vacuum systems and protect the laboratory. Whatman™ Vacu-Guard Filter protects vacuum pump systems from solvent vapor or gaseous contaminants and aqueous aerosols.
  • Excellent for protecting vacuum pump systems from solvent vapor or gaseous contaminants and aqueous aerosols
  • Designed for in-line use with stepped barb connections for 10 to 12mm ID hose
  • Membrane retains 99.99% of airbone particles greaterthan 0.1μm
  • Features hydrophobic PTFE membrane
Specifications
Material (Membrane)PTFE
Pore Size0.45 μm
Filtration Area16 cm2
For Use With (Application)Isolate infectious materials
Housing MaterialPolypropylene
Inlet Style6 to 10 mm (1/4 to 3/8 in.) stepped barb
Outlet Style6 to 10 mm (1/4 to 3/8 in.) stepped barb
Product LineVacu-Guard Filter
Quantity10Pack
TypeInline Filter
Diameter (Metric)50 mm
Diameter (Metric) Disc50 mm
Unit SizePK