SP Scienceware™ Sterileware™ Sterile Mini-Tongs
SP Scienceware™ Sterileware™ Sterile Mini-Tongs
Actual product may vary
Fisher Catalogue
SP Scienceware™ Sterileware™ Sterile Mini-Tongs
These small, lightweight tongs are ideal for sampling small quantities of material ranging from food to pharmaceuticals
Have Questions?
Catalog number FKS11362164
View Price:Sign in to see your account pricing. Need an account? Register with us today.
These small, lightweight tongs are ideal for sampling small quantities of material ranging from food to pharmaceuticals
  • Ridged jaws securely hold wet or dry samples
  • One piece, sterile tongs are made from polystyrene plastic
  • Individually packaged, sterilized by gamma radiation, lot stamped for accurate lot tracing and shipped with a Certificate of Processing for Sterilization
Specifications
For Use With (Application)Ideal for sampling small quantities of material ranging from food to pharmaceuticals
Length (English)4.25 in.
Tip StyleSerrated
Length (Metric)11 cm
MaterialPolystyrene
TypeMini Tongs
Unit SizePK