Quickfit™ Dreschel Bottle Head for Gas Washing Bottle
Quickfit™ Dreschel Bottle Head for Gas Washing Bottle
Actual product may vary
Fisher Catalogue
Quickfit™ Dreschel Bottle Head for Gas Washing Bottle
Complies with BS 2461. Quickfit™ Dreschel Bottle Head for Gas Washing Bottle includes detachable plastic connectors for easy fitting of 9mm bore flexible tubing. Bottle head and gas washing bottle should be ordered separately.
Have Questions?
Catalog NumberLength (Metric)For Use With (Equipment)
FKS11362055Stem: 130mmMF29/3/125
Catalog number FKS11362055
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Length (Metric):
Stem: 130mm
For Use With (Equipment):
MF29/3/125
Complies with BS 2461. Quickfit™ Dreschel Bottle Head for Gas Washing Bottle includes detachable plastic connectors for easy fitting of 9mm bore flexible tubing. Bottle head and gas washing bottle should be ordered separately.
  • Complies with BS 2461
  • Dreschel type
  • With detachable plastic connectors for easy fitting of 9mm bore flexible tubing
Specifications
ColorClear
IncludesDetachable plastic connectors
MaterialBorosilicate Glass
Length (Metric)Stem: 130mm
Quantity1PK
Certifications/ComplianceBS 2461
ShapeCylindrical
Product TypeDreschel Bottle Head
For Use With (Equipment)MF29/3/125
Length (English)5.11 in.
Through the Closure DispensingYes
Unit SizeEA