Check Order Status
Filter Holder
Actual product may vary
Fisherbrand
Filter Holder
Polysulfone In-Line Filter Holder for 50mm Diameter Membrane
Have Questions?
Catalog Number
Material
Quantity
FKS11359304
Polysulfone
10 Pk.
Catalog number
FKS11359304
Price (SGD)
442.00
PK
Add to cart
Material:
Polysulfone
Quantity:
10 Pk.
Price (SGD)
442.00
PK
Add to cart
Polysulfone In-Line Filter Holder for 50mm Diameter Membrane
Specifications
Material
Polysulfone
Model
FP050/1 PSU
Quantity
10 Pk.
Type
Filter Holder
Diameter (Metric) Membrane
50 mm