Amber Glass Wide Neck Powder Bottles with Black Urea Screw Caps
Amber Glass Wide Neck Powder Bottles with Black Urea Screw Caps
Actual product may vary
Fisherbrand
Amber Glass Wide Neck Powder Bottles with Black Urea Screw Caps
Use these bottles for general storage applications. Azpack™ Amber Glass Wide Neck Powder Bottles with Black Urea Screw Caps provide protection for light sensitive liquids.
Have Questions?
Catalog NumberCapacity (Metric)Closure Size
FKS1135802315 mL28/R3
Catalog number FKS11358023
Price (SGD)
153.00
PK
Add to cart
Capacity (Metric):
15 mL
Closure Size:
28/R3
Price (SGD)
153.00
PK
Add to cart
  • Constructed of amber-colored soda lime glass
  • Includes black urea screw cap closure
  • Closure lined with wood pulp faced steran (PVDC)
Specifications
Capacity (Metric)15 mL
ColorAmber
MaterialSoda Lime Glass
Quantity40/PK
Closure ColorBlack
Closure IncludedYes
MouthWide
Height (Metric)55 mm
Closure LinerWood Pulp-faced PVDC
Closure MaterialUrea
Closure Size28/R3
Diameter (Metric)33 mm
Closure TypeScrew Cap
Unit SizePK