Premium Microscope Slide Box with Cork Lining
Premium Microscope Slide Box with Cork Lining
Actual product may vary
Fisherbrand
Premium Microscope Slide Box with Cork Lining
Store and transport slides safely with durable ABS cases. Fisherbrand™ Colored ABS Slide Boxes keep standard microscope slides in numbered, grooved slots. Provides safe, dust-free storage.
Have Questions?
Catalog NumberColor
FKS11354135White
Catalog number FKS11354135
Price (SGD)
32.00
EA
Add to cart
Color:
White
Price (SGD)
32.00
EA
Add to cart
  • Molded, rigid boxes securely hold slides in grooved slots
  • Both the lining and the slots are numbered to correspond to the slide inventory sheet
  • Friction-fit cover closes easily and allows for easy access to slides
  • Features a thumb-latch lock mechanism for additional security
  • Stackable
Specifications
Slide Capacity25
Closure TypeFriction-Fit Lid
ColorWhite
Device Holds25 Slides
Dimensions (L x W x H)141 x 88 x 35 mm
For Use With (Application)Safely storing or transporting microscope slides
For Use With (Equipment)Standard 25 x 75 mm (1 x 3 in.) Microscope Slides
Height (English)1.37 in.
Height (Metric)35 mm
IncludesSlide Inventory Sheet
Length (English)5.55 in.
Length (English) Slide1 in.
Length (Metric)141 mm
Length (Metric) Slide25 mm
MaterialABS
ShapeRectangular
StackableYes
SterilityNon-sterile
Width (English)3.46 in.
Width (English) Slide3 in.
Width (Metric)88 mm
Width (Metric) Slide75 mm
Lining MaterialCork
Unit SizeEA