Azlon™ Polypropylene Squat Form Cylinder
Actual product may vary
Fisherbrand
Azlon™ Polypropylene Squat Form Cylinder
Manufactured from high clarity polypropylene for good chemical and temperature resistance. The design of Azlon™ Polypropylene Squat Form Cylinders provides increased stability on the bench and ease of handling. Ideal for use in lab areas with restricted space.
Have Questions?
Catalog NumberCapacity (Metric)Graduations
FKS113534341000 mL20 mL
Catalog number FKS11353434
Price (SGD)
83.00
Each
Capacity (Metric):
1000 mL
Graduations:
20 mL
Price (SGD)
83.00
Each
Design Features:
  • Squat design ensures stability
  • Unique self-draining base eliminates water collection during washing in an automatic laboratory washing machine resulting in the elimination of spill and contamination
  • Molded graduations with blue over-print provide durability and visibility for measurement
  • Anti-suction base minimizes spillage when lifting cylinder from wet surface
Specifications
Capacity (Metric)1000 mL
MaterialPolypropylene
ClassClass B
Scaleblue
Diameter (Metric)83 mm
Quantity1/Pk.
Graduations20 mL
Scale ColorBlue
Base TypeCircular
Height (Metric)285 mm
Product TypeUngraduated Measuring Cylinder
Unit SizeEach