SP Scienceware™ Holdfast™ Polypropylene Graduated Cylinders
SP Scienceware™ Holdfast™ Polypropylene Graduated Cylinders
Actual product may vary
Fisherbrand
SP Scienceware™ Holdfast™ Polypropylene Graduated Cylinders
Prevents tipping and rolling due to hexagonal base. Bel Art™ SP Scienceware™ Holdfast™ Polypropylene Graduated Cylinders include handle for a secure, solid grip for safer pouring.
Have Questions?
Catalog NumberCapacity (Metric)Graduations
FKS113521642000 mL20 mL
Catalog number FKS11352164
Price (SGD)
339.00
EA
Add to cart
Capacity (Metric):
2000 mL
Graduations:
20 mL
Price (SGD)
339.00
EA
Add to cart
Prevents tipping and rolling due to hexagonal base. Bel Art™ SP Scienceware™ Holdfast™ Polypropylene Graduated Cylinders include handle for a secure, solid grip for safer pouring.
  • One-piece cylinder with handle provides a secure, solid grip for safer pouring
  • Easy-to-read double scale with raised graduations
  • Hexagonal base prevents tipping and rolling
  • Will not create a meniscus
  • Handle opening: 8.9 × 2.9cm (3 × 1.20 in.)
  • Chemically resistant polypropylene
  • Autoclavable at 121°C (250°F)
Specifications
Capacity (Metric)2000 mL
MaterialPolypropylene
CalibrationTo Contain, To Deliver
Diameter (English)3.5 in.
ScaleDual
IncludesLarge Handle
Graduations20 mL
AutoclavableAutoclavable
Subdivision20mL
ShapeRound
Base TypeHexagonal
Height (English)19.3 in.
Height (Metric)490.2 mm
TypeGraduated Cylinder
Unit SizeEA