Check Order Status
Polypropylene Screw Lid GL45 Kit
Actual product may vary
Fisherbrand
Polypropylene Screw Lid GL45 Kit
Used to secure containers. Cytiva Polypropylene Screw lid GL45 Kit comes with screw lid and cap membrane.
Have Questions?
Catalog Number
Material
FKS11350192
Polypropylene, Ethylenepropylenediene
Catalog number
FKS11350192
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Material:
Polypropylene, Ethylenepropylenediene
The kit contains 1 x Screw lid 1 x Cap membrane
The membrane should not be used when working with organic solvent.
Lid: Polypropylene (PP)
Membrane: Ethylenepropylenediene (EPDM)
Specifications
For Use With (Equipment)
Lid
Material
Polypropylene, Ethylenepropylenediene
Type
Screw Lid GL45 Kit
Includes
Screw lid, Cap membrane
Quantity
1 Kit