Search Thermo Fisher Scientific
Catalog Number | Volume (Metric) | Graduations | Tolerance |
---|---|---|---|
FKS11349663 | 25 mL | 0.1 mL | ±0.1 mL |
FKS11339663 | 10 mL | 0.1 mL | ±0.05 mL |
FKS11329663 | 5 mL | 0.05 mL | ±0.03 mL |
FKS11309663 | 1 mL | 0.01 mL | ±0.007 mL |
FKS11319663 | 2 mL | 0.02 mL | ±0.01 mL |