Pyrex™ Aspirator Bottle with Ground Glass Side Socket
Pyrex™ Aspirator Bottle with Ground Glass Side Socket
Actual product may vary
Fisherbrand
Pyrex™ Aspirator Bottle with Ground Glass Side Socket
Manufactured from borosilicate glass
Have Questions?
Catalog NumberCapacity (Metric)
FKS113478845000 mL
Catalog number FKS11347884
Price (SGD)
712.00
EA
Add to cart
Capacity (Metric):
5000 mL
Price (SGD)
712.00
EA
Add to cart
Manufactured from borosilicate glass
  • Ground side socket (size 29/32) to accept stopcock assemblies
  • Unground top neck to accept rubber stoppers
Specifications
Capacity (Metric)5000 mL
Diameter (Metric) Inner Neck44 mm
MaterialBorosilicate Glass
Height (Metric)318 mm
Product TypeAspirator Bottle
Diameter (Metric)181 mm
Unit SizeEA