Samco™ Fine Tip Transfer Pipettes
Samco™ Fine Tip Transfer Pipettes
Actual product may vary
Samco™ Fine Tip Transfer Pipettes
Samco™ Fine Tip Transfer Pipettes
Samco™ Fine Tip Transfer Pipettes
Samco™ Fine Tip Transfer Pipettes
Samco™ Fine Tip Transfer Pipettes
Samco™ Fine Tip Transfer Pipettes
Samco™ Fine Tip Transfer Pipettes
Samco™ Fine Tip Transfer Pipettes
Samco™ Fine Tip Transfer Pipettes
Samco™ Fine Tip Transfer Pipettes
Samco™ Fine Tip Transfer Pipettes
Samco™ Fine Tip Transfer Pipettes
Samco™ Fine Tip Transfer Pipettes
Samco™ Fine Tip Transfer Pipettes
Samco™ Fine Tip Transfer Pipettes
Samco™ Fine Tip Transfer Pipettes
Samco™ Fine Tip Transfer Pipettes
Samco™ Fine Tip Transfer Pipettes
Samco™ Fine Tip Transfer Pipettes
Samco™ Fine Tip Transfer Pipettes
Samco™ Fine Tip Transfer Pipettes
Samco™ Fine Tip Transfer Pipettes
Samco™ Fine Tip Transfer Pipettes
Samco™ Fine Tip Transfer Pipettes
Samco™ Fine Tip Transfer Pipettes
Samco™ Fine Tip Transfer Pipettes
Samco™ Fine Tip Transfer Pipettes
Samco™ Fine Tip Transfer Pipettes
Samco™ Fine Tip Transfer Pipettes
Fisherbrand
Samco™ Fine Tip Transfer Pipettes
Perform precise dispensing of small drops into small-volume containers using these fine-tip transfer pipettes.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)SterilityBulb Draw (Metric)Packaging Type
FKS113478635.0 mLSterile3.1 mL20/bag, 400/pk
FIS13-711-251.7 mLNon-sterile1.0 mLBulk 400/Pk 8Pk/Cs 3200/Cs
FIS13-711-295.4 mLNon-sterile3.1 mLBulk 250/Pk 8Pk/Cs 2000/Cs
FIS13-711-265.8 mLNon-sterile3.3 mL500/Pk., 5000/Cs.
FIS13-711-275.8 mLSterile3.3 mLIndividually Wrapped
FIS13-711-285.8 mLSterile3.3 mL20/Bag, 500/Pk., 4000/Cs.
FIS13-711-308.9 mLNon-sterile4.8 mLBulk 400/Pk 10Pk/Cs 4000/Cs
Catalog number FKS11347863
Price (SGD)
175.00
CS
Add to cart
Volume (Metric):
5.0 mL
Sterility:
Sterile
Bulb Draw (Metric):
3.1 mL
Packaging Type:
20/bag, 400/pk
Price (SGD)
175.00
CS
Add to cart
These fine-tip transfer pipettes are ideal for removing separating liquid samples. Designed with a built-in pipette bulb, they allow precise dispensing of small drops into small-volume containers.
  • Made of shatterproof, nontoxic polyethylene
  • Low-affinity surface – prevents proteins binding in research applications
  • Can be frozen in liquid nitrogen, or heat sealed for storage
  • Available with fine tip or extended fine tip
Specifications
Bulb Draw (Metric)3.1 mL
DescriptionSamco 5.0mL Extended Fine Tip Transfer Pipets, Large Bulb, Sterile, 20/pk, White Box
Diameter (Metric) Outer3.0 mm
MaterialLDPE
SterilitySterile
GraduatedNo
Packaging Type20/bag, 400/pk
Volume (Metric)5.0 mL
Length (Metric)15.3 cm
TypeExtended Fine Tip Transfer Pipet
Unit SizeCS