1x1 image pixel for data collection
Mechanical Benchtop Counter
Mechanical Benchtop Counter
Actual product may vary
Fisherbrand
Mechanical Benchtop Counter
Depress the keys in the appropriate blood cell category to keep track of counts. Mechanical Benchtop Counter is an economical solution for counting blood cells, bacterial colonies and other types of repetitive counting.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescriptionLength (Metric)
FKS113440959-key Counter320 mm
FKS113340956-key Counter250 mm
Catalog number FKS11344095
Price (SGD)
1,394.00
EA
Add to cart
Description:
9-key Counter
Length (Metric):
320 mm
Price (SGD)
1,394.00
EA
Add to cart
  • Each key records up to 999
  • Total window keeps track of total number of strokes made on all other keys
  • Bell rings when total reaches 100 blood cell key labels
    • HS5360A:
      • Baso., Eosin., Mono., Lymph., Poly., and Total,
      • Measures 250 x 76 x 56 mm
    • HS5360B:
      • Baso, Eosin., Myelo., Juven., Stab., Seg., Lymph., Mono., and Total,
      • Measures 320 x 76 x 56 mm
Specifications
Product TypeMechanical Counter
Display TypeMechanical
Description9-key Counter
ModelHS5360B
Quantity1 Ea.
Height (English)2.2 in.
Height (Metric)56 mm
Length (English)12.6 in.
Length (Metric)320 mm
Width (English)3 in.
Width (Metric)76 mm
IncludesLabels: Baso, Eosin., Myelo., Juven., Stab., Seg., Lymph., Mono., and Total
Unit SizeEA
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link