Whatman™ Antibiotic Assay Discs
Whatman™ Antibiotic Assay Discs
Actual product may vary
Whatman™ Antibiotic Assay Discs
Used in conjunction with the Kirby-Bauer method of antibiotic testing. Whatman™ Antibiotic Assay Discs determine the type of causal agent of infectious diseases and their sensitivity to antibiotics and chemotherapeutic agents.
Have Questions?
Catalog NumberDiameter (Metric)
FKS113406546 mm
Catalog number FKS11340654
Price (SGD)
874.00
PK
Add to cart
Diameter (Metric):
6 mm
Price (SGD)
874.00
PK
Add to cart
Used in conjunction with the Kirby-Bauer method of antibiotic testing. Whatman™ Antibiotic Assay Discs determine the type of causal agent of infectious diseases and their sensitivity to antibiotics and chemotherapeutic agents.
Antibiotic assay discs are used for determining the type of causal agent of infectious diseases and for checking their sensitivity to antibiotics and chemotherapeutic agents in vitro by means of the inhibition zone determination method.
  • For use with Kirby-Bauer method of antibiotic testing
  • Allows rational and selective chemotherapy
Specifications
Product TypeAntibiotic Assay Discs
Test TypeInfectious Disease, Antibiotic Susceptibility Sensitivity
Packaging Type1000 Discs/Pk.
Quantity1000 Discs/Pk.
Diameter (Metric)6 mm
Unit SizePK