1x1 image pixel for data collection
Borosilicate Glass Aspirator Stopcock with Ground Cone
Borosilicate Glass Aspirator Stopcock with Ground Cone
Actual product may vary
Fisherbrand
Borosilicate Glass Aspirator Stopcock with Ground Cone
Inserts into the ground glass side-socket of an aspirator botte. Pyrex™ Borosilicate Glass Aspirator Stopcock with Ground Cone provides a leakproof seal.
Have Questions?
Catalog NumberIncludesLength (Metric)
FKS11337894Traditional Glass key95 mm
Catalog number FKS11337894
Price (SGD)
337.00
EA
Estimated availability date 01-Oct-2025
Add to cart
Includes:
Traditional Glass key
Length (Metric):
95 mm
Price (SGD)
337.00
EA
Add to cart
Inserts into the ground glass side-socket of an aspirator botte. Pyrex™ Borosilicate Glass Aspirator Stopcock with Ground Cone provides a leakproof seal.
  • For use with aspirator bottles 1541 and 1569
  • Cone size: 29/32
  • Type 1568/04M with Rotaflo™ PTFE key which is chemically inert and requires no lubrication
  • Type 1571/02M with traditional glass key
Specifications
TypeAspirator Stopcock
Plug Bore4 mm
MaterialBorosilicate Glass
Length (Metric)95 mm
For Use With (Equipment)Aspirator bottles 1541 and 1569
IncludesTraditional Glass key
Unit SizeEA
Documents & Downloads
Certificates
0 results displayed, search above for a specific certificate
Share catalog number, name or link