Carton Biohazard Incineration
Carton Biohazard Incineration
Actual product may vary
Fisherbrand
Carton Biohazard Incineration
Perfect for containing biohazardous waste slated for incineration. Bel Art™ Carton Biohazard Incineration cartons can be incinerated by placing the entire unit in the incinerator.
Have Questions?
Catalog NumberHeight (Metric)
FKS11332204250 mm
Catalog number FKS11332204
Price (SGD)
347.00
PK
Add to cart
Height (Metric):
250 mm
Price (SGD)
347.00
PK
Add to cart
  • Tabs along the top edge keep a clear 0.038mm (1.5 mil) thick polypropylene biohazard bag from slipping out of place
  • Snug-fitting lid has a flap for easily inserting waste material
  • Printed with large, bright, biohazard graphics and instructions in four languages (English, Spanish, French, German)
  • Packaged flat to save space and shipping costs; carton and inner bag can be assembled in seconds
  • Available in sizes to suit benchtop or floor use
  • Optional, reusable, sanitary cover (F13204-0000) for floor model sold separately
Specifications
Height (Metric)250 mm
Thickness0.04 mm
Product TypeCarton Biohazard Incineration
Unit SizePK