Sticky Tape
Sticky Tape
Actual product may vary
Fisher Catalogue
Sticky Tape
Alternative to flask clamps for securing vessels to a shaker platform
Have Questions?
Catalog number FKS11325974
View Price:Sign in to see your account pricing. Need an account? Register with us today.
  • Ideal for containers such as bottles and dishes that cannot be held with conventional flask clamps
  • With flasks, tape can be used at speeds of up to 200 to 300 rpm depending on orbit diameter, flask size and load
  • Roll size: 5m L × 4cm W (16.4 ft. × 1.6 in.)
  • Can be used several times before losing its adhesive properties
  • Cannot be used in water baths
Specifications
Product TypeSticky Tape
Dimensions (L x W)16.4 ft. x 1.6 in. (5 m x 4 cm)
Length (English)16.4 ft.
Length (Metric)499 cm
Width (English)1.6 in.
Width (Metric)4 cm
For Use With (Application)Holding flasks and other vessels to platform of orbital shaker
Unit SizeEA