Pyrex™ Graduated Dropping Funnel with Glass Key
Pyrex™ Graduated Dropping Funnel with Glass Key
Actual product may vary
Fisherbrand
Pyrex™ Graduated Dropping Funnel with Glass Key
Used to transfer fluids.
Have Questions?
Catalog NumberCapacity (Metric)
FKS11313684250 mL
Catalog number FKS11313684
Price (SGD)
556.00
EA
Add to cart
Capacity (Metric):
250 mL
Price (SGD)
556.00
EA
Add to cart
  • Complies with ISO 4800 type 4
  • Manufactured from Pyrex borosilicate glass
  • Double graduated scale showing approximate volume contained/dispensed
  • Graduations in white enamel
  • With interchangeable glass key
  • Supplied with glass stopper
Specifications
Capacity (Metric)250 mL
MaterialBorosilicate Glass, Borosilicate Glass
GraduatedYes
IncludesGlass stopper and key
Graduations5mL
Product TypeDropping Funnel
Unit SizeEA