Quickfit™ Vacuum Receiver Adapter
Quickfit™ Vacuum Receiver Adapter
Actual product may vary
Fisherbrand
Quickfit™ Vacuum Receiver Adapter
Deliver liquids between glass containers under vacuum. QuickFit™ vacuum receiver adapter comes with a side arm for easy attachement to vacuum supply.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberHeight (Metric)Cone SizeStandard Taper Joint No.
FKS11312105165mm29/3224/29
FKS11302105150mm24/2924/29
Catalog number FKS11312105
Price (SGD)
373.00
EA
Add to cart
Height (Metric):
165mm
Cone Size:
29/32
Standard Taper Joint No.:
24/29
Price (SGD)
373.00
EA
Add to cart
Deliver liquids between glass containers under vacuum. QuickFit™ vacuum receiver adapter comes with a side arm for easy attachement to vacuum supply.
  • Delivery stem within cone
  • With sidearm for connection to a vacuum supply
  • Easy connection to flexible tubing via detachable plastic screwthread connector
Specifications
Cone Size29/32
For Use With (Equipment)Flexible tubing
Height (English)6.49in.
Height (Metric)165mm
IncludesDetachable Plastic screw thread connector, delivery stem, side-arm
MaterialBorosilicate Glass
Product TypeVacuum receiver angled adapter
Standard Taper Joint No.24/29
Unit SizeEA