Check Order Status
Amber Borosilicate Glass Class A Volumetric Flask with Stopper
Actual product may vary
Fisher Catalogue
Amber Borosilicate Glass Class A Volumetric Flask with Stopper
Amber Borosilicate Glass Volumetric Flask includes interchangeable polyethylene stopper.
Have Questions?
Catalog Number
Capacity (Metric)
Closure Size
Tolerance
FKS11309443
50 mL
12/21
±0.06 mL
Catalog number
FKS11309443
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Capacity (Metric):
50 mL
Closure Size:
12/21
Tolerance:
±0.06 mL
Amber Borosilicate Glass Volumetric Flask includes interchangeable polyethylene stopper.
With one mark graduation and interchangeable polyethylene stopper.
The details on each flask include confirmation of compliance to International Standards as well as an individual re-order code.
The Fisherbrand 'Batch Certificate of Compliance' is downloadable for this item from the 'Documentation' tab
Specifications
Certifications/Compliance
DIN 12691
Closure Material
Polypropylene
Color
Amber
Capacity (Metric)
50 mL
Material
Borosilicate Glass
Height (Metric)
139 mm
No. per Case
1 Each
Tolerance
±0.06 mL
Graduated
Yes
Class
A
Closure Type
Cap
Diameter (Metric) Outer
49 mm
Closure Size
12/21