Thick Biohazard Disposal Bags, 1.5 mil
Thick Biohazard Disposal Bags, 1.5 mil
Actual product may vary
Thick Biohazard Disposal Bags, 1.5 mil
Thick Biohazard Disposal Bags, 1.5 mil
Thick Biohazard Disposal Bags, 1.5 mil
Fisherbrand
Thick Biohazard Disposal Bags, 1.5 mil
Include black biohazard symbol and precautionary procedures printed in four languages. Bags are autoclavable and are effective for the disposal of biohazardous materials.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (English)
FKS113021645 to 9 gal.
FIS03-411-7002 to 4 gal.
FIS03-411-7015 to 9 gal.
FIS03-411-70213 to 20 gal.
Catalog number FKS11302164
Price (SGD)
1,077.00
PK
Add to cart
Capacity (English):
5 to 9 gal.
Price (SGD)
1,077.00
PK
Add to cart
Include black biohazard symbol and precautionary procedures printed in four languages. Bags are autoclavable and are effective for the disposal of biohazardous materials.
  • Bright red polypropylene bags include black biohazard symbol
  • Precautionary procedures printed on bag in English, Spanish, French, and German for added safety
  • Sterilization patch darkens when subjected to steam sterilization
  • High temperature steam autoclavable to 135°C (275°F)
Specifications
Description19W x 23H in. (48 x 58cm)
AutoclavableAutoclavable
Capacity (English)5 to 9 gal.
Capacity (Metric)26 L
Certifications/ComplianceASTM 1922 Tear Resistance, ASTM 1709 Dart Impact
Closure TypeZip Tie
ColorRed
DisposableSingle-use
For Use With (Application)Used for biohazard waste
IncludesPre-printed sterilization indicator patch, labeled with the standard biohazard warning symbol or precautionary protocols English, French, German, Spanish
MaterialPolypropylene
SterilityNon-sterile
Thickness (Metric)0.279 mm
Thickness (English)1.50 mil
Dimensions (L x W)19 x 23 in.
Length (English)23 in.
Length (Metric)58 cm
Width (English)19 in.
Width (Metric)48 cm
Height (English)23 in.
Height (Metric)58 cm
Product TypeBiohazard Disposal Bag
Unit SizePK