Check Order Status
Heating Mantle
Actual product may vary
Fisherbrand
Heating Mantle
Use these heating elements with built in controllers for a variety of applications. Fisherbrand™ Heating Mantles are metal cased with a unique airflow keeping the exterior cool to the touch.
Have Questions?
Change view
Catalog Number
Capacity (Metric)
FKS10884191
250 mL
FKS10332290
50 mL
2 Options
Catalog number
FKS10884191
Price (SGD)
1,799.00
EA
Add to cart
Capacity (Metric):
250 mL
Price (SGD)
1,799.00
EA
Add to cart
Doubled fused and earthed screen
Heating element cartridges are replaceable
No external controllers are needed
'Heater On' and 'Power-On' lights
Maximum element temperature of 450°C
Specifications
Controller Type
Integrated
Capacity (Metric)
250 mL
Voltage
230 V
Electrical Requirements
230V
Includes
Controller
Material
Metal cased
Shape
Round
Wattage
150w
Product Type
Heating Mantle
Temperature (Metric)
450°C
Frequency
50 Hz