Quickfit™ Borosilicate Glass Short Neck Round Bottom Flask
Quickfit™ Borosilicate Glass Short Neck Round Bottom Flask
Actual product may vary
Quickfit
Quickfit™ Borosilicate Glass Short Neck Round Bottom Flask
Use these flasks for a variety of applications. Quickfit™ Borosilicate Glass Short Neck Round Bottom Flask are manufactured from borosilicate glass for corrosion-free performance and resistance to thermal shock.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Socket Joint
FKS1080990050 mL14/23
FKS104885601000 mL29/32
FKS10829580100 mL14/23
FKS10538190100 mL24/29
FKS10671071150 mL24/29
FKS1076511125 mL14/23
FKS1055859025 mL24/29
FKS12682002250 mL24/29
FKS10488570500 mL24/29
FKS1018178050 mL24/29
Catalog number FKS10809900
Price (SGD)
367.00
PK
Add to cart
Capacity (Metric):
50 mL
Socket Joint:
14/23
Price (SGD)
367.00
PK
Add to cart
Use these flasks for a variety of applications. Quickfit™ Borosilicate Glass Short Neck Round Bottom Flask are manufactured from borosilicate glass for corrosion-free performance and resistance to thermal shock.
  • Hgh quality Quickfit™ ground glass socket
  • Comprehensive range of 46 flasks from 5mL to 10,000mL
  • Body diameter and wall thickness comply with relevant ISO 1773 and ISO 4797 requirements
Specifications
Certifications/ComplianceISO 1773, ISO 4797
Capacity (Metric)50 mL
MaterialBorosilicate Glass
Height (Metric)90 mm
Neck StyleShort
Socket Joint14/23
Bottom ShapeRound
Diameter (Metric) Outer50 mm
Unit SizePK