Varipette™ 4720 Pipettes
Varipette™ 4720 Pipettes
Actual product may vary
Fisherbrand
Varipette™ 4720 Pipettes
Use this pipette for liquids with a high vapour pressure or viscosity. Eppendorf™ Varipette™ 4720 Pipette is a continuously adjustable pipettor for precise pipetting of volumes up to 10mL in 10μL increments.
Have Questions?
Catalog NumberVolume (Metric)
FKS107975911 to 10 μL
Catalog number FKS10797591
Price (SGD)
3,213.00
EA
Add to cart
Volume (Metric):
1 to 10 μL
Price (SGD)
3,213.00
EA
Add to cart
Varitip P can be used for aspirating 1mL to 10mL from beakers according to the positive displacement principle. Varitip S is used for aspirating 2. 5mL to 10mL from tall, narrow neck vessels. System consists of a dispensing part and the Maxitip air displacement principle. No carry over as Maxitip exchanged. Valve for Maxitip ensures drip free dispensing of liquids with a high vapour pressure. Graduated Maxitip G for compensation of nominal value of Varipette™.
Specifications
Product LineVaripette 4720
TypeAdjustable Volume Pipette
Volume (Metric)1 to 10 μL
For Use With (Application)Used for Accurate Pipetting of Viscous Solutions or of Liquids with High-Vapor Pressure
Functional Range1 to 10mL
Quantity1 Each
Unit SizeEA