Glass Fiber Nylon Spatula, with spoon ends
Glass Fiber Nylon Spatula, with spoon ends
Actual product may vary
Fisher Catalogue
Glass Fiber Nylon Spatula, with spoon ends
Manufactured from glass fiber nylon for excellent chemical resistance. Kartell™ Glass Fiber Nylon Spatulas are suitable for use in food and beverage industry, laboratory work and educational settings.
Have Questions?
Catalog NumberLength (Metric)
FKS10796572210 mm
Catalog number FKS10796572
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Length (Metric):
210 mm
Manufactured from glass fiber nylon for excellent chemical resistance. Kartell™ Glass Fiber Nylon Spatulas are suitable for use in food and beverage industry, laboratory work and educational settings.
  • General purpose spatulas with excellent chemical resistance
  • Scraper plus spoon or double scraper configuration
  • Autoclavable
  • Manufactured to DIN12890 standard
Specifications
Quantity10 Pk.
AutoclavableAutoclavable
ShapeSpoon
Capacity (Metric)1.8 mL
ColorRed
Certifications/ComplianceNSF/ANSI Standard 51, DIN 12890
Product TypeGeneral Purpose Spatula
For Use With (Application)For use in food and beverage industry, inexpensive general purpose spatulas
Width (Metric)15 to 20 mm
MaterialGlass Fiber Nylon
Length (Metric)210 mm
Unit SizePK