HDPE Pasteur Pipet
HDPE Pasteur Pipet
Actual product may vary
Fisherbrand
HDPE Pasteur Pipet
Use these pipets for simple and safe liquid handling. Ratiolab™ HDPE Pasteur Pipets feature a low affinity surface which reduces the loss of cells and valuable proteins through binding.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)Length (Metric)Sterility
FKS107898231 mL150 mmSterile
FKS102013511 mL150 mmNon-sterile
FKS101781113 mL155 mmNon-sterile
Catalog number FKS10789823
Price (SGD)
164.00
PK
Add to cart
Volume (Metric):
1 mL
Length (Metric):
150 mm
Sterility:
Sterile
Price (SGD)
164.00
PK
Add to cart
  • No risk of cross-contamination or carry-over
  • No danger of glass breakage with the resulting danger of infectious material escaping
  • Transparent polyethylene construction
  • Inert to biological fluids and most acids
  • Soft, elastic bulb
  • Burr-free orifice for neat, clean and precise dispensing
  • BSE-free
  • Can be used for food
Specifications
MaterialHDPE
SterilitySterile
For Use With (Application)Food analysis
Quantity500 Pack
Volume (Metric)1 mL
Length (Metric)150 mm
TypePasteur Pipette
Unit SizePK