Sterile Spatulas, Tapered blade/spoon
Sterile Spatulas, Tapered blade/spoon
Actual product may vary
Fisher Catalogue
Sterile Spatulas, Tapered blade/spoon
Constructed of rigid nontoxic polystyrene to provide good chemical resistance to aqueous solutions. Corning™Sterile Spatulas are an ideal solution for molecular biology applications, lyophilized samples, and contamination-free sampling.
Have Questions?
Catalog NumberLength (Metric)
FKS1078801224.9 cm
Catalog number FKS10788012
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Length (Metric):
24.9 cm
Constructed of rigid nontoxic polystyrene to provide good chemical resistance to aqueous solutions. Corning™Sterile Spatulas are an ideal solution for molecular biology applications, lyophilized samples, and contamination-free sampling.
  • Corning spatulas are designed to save time and to provide contamination-free samples.
  • Eliminates the recycling and resterilizing necessary with reusable spatulas.
  • Available in five different configurations.
  • Microspatulas available in two configurations.
  • Individually packaged.
  • Antistatic.
  • Sterile.
  • RNase-/DNase-free.
  • Nonpyrogenic.
Specifications
SterilitySterile
ShapeTapered Flat End, Spoon End
Certifications/ComplianceUSP Class VI, non-pyrogenic, RNase and DNase-free Certified
DisposableSingle-use
Product TypeSpatula
For Use With (Application)Molecular biology applications, lyophilized samples and contamination-free sampling
Length (English)9.8 in.
IncludesTapered flat blade at one end and a spoon at the other end
MaterialPolystyrene
Handle TypeTapered
Length (Metric)24.9 cm
Unit SizePK