Thermo Scientific™ Stainless Steel Dressing Scissors
Thermo Scientific™ Stainless Steel Dressing Scissors
Actual product may vary
Fisherbrand
Thermo Scientific™ Stainless Steel Dressing Scissors
Manufactured from stainless steel. Bochem™ Thermo Scientific™ Stainless Steel Dressing Scissors comes in three types.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (Metric)Tip Style
FKS10787191155 mmSharp/Round
FKS10088360130 mmSharp/Sharp
Catalog number FKS10787191
Price (SGD)
27.00
PK
Add to cart
Length (Metric):
155 mm
Tip Style:
Sharp/Round
Price (SGD)
27.00
PK
Add to cart
  • Three types: round-round, sharp-round, and sharp-sharp
  • Stainless steel
Specifications
Product TypeDressing Scissors
Length (Metric)155 mm
MaterialStainless Steel
Tip StyleSharp/Round
Unit SizePK