PTFE Anchor Propeller Stirrer Shaft
PTFE Anchor Propeller Stirrer Shaft
Actual product may vary
Fisherbrand
PTFE Anchor Propeller Stirrer Shaft
Use with shaft guides for mixing high viscosity substances at slower speeds. The PTFE anchor stir shafts have a stainless-steel core.
Have Questions?
Catalog NumberLength (Metric)Diameter (Metric) ShaftDiameter (Metric) Fan
FKS10780333550 mm10 mm100 mm
Catalog number FKS10780333
Price (SGD)
248.00
EA
Add to cart
Length (Metric):
550 mm
Diameter (Metric) Shaft:
10 mm
Diameter (Metric) Fan:
100 mm
Price (SGD)
248.00
EA
Add to cart
PTFE shaft stirrers have a unique, fully molded construction featuring a rotor molded to a fully PTFE-encapsulated steel reinforced drive shaft.
  • Chemically resistant, inert, and leak-free
  • Easy to clean
  • No cross contamination
  • Shatterproof
  • Non-scratch
  • Use in temperatures up to approximately 280°C
  • Exposed shaft models (EX) are produced with a stainless steel core with an exposed end for more rigid clamping to the chuck drive
  • For mixing high viscosity substances at slower speeds
Specifications
Diameter (Metric) Fan100 mm
Diameter (Metric) Shaft10 mm
For Use With (Equipment)Universal Stirrer Guide
For Use With (Application)Mixing
Length (Metric)550 mm
Quantity1 Each
Temperature (Metric)-200°C to +280°C
TypePTFE Shaft Stirrer Anchor
Unit SizeEA